Would you like to log in again?
Automation Tips for Analytik Jena Cybio Felix, Well Vario and Selma Workstations
Supplier: Fluotics
CYBW-25Q.NS CYBW-25Q.F CYBW-25.NS CYBF-250.ST CYBW-10Q.ST CYBF-50.ST CYBW-10.ST CYBW-25Q.ST CYBW-25.ST CYBF-50.NS CYBW-25.F CYBF-50.F CYBW-60Q.F CYBW-10.NS CYBW-60Q.ST CYBF-250.NS CYBW-60.NS CYBW-60.ST CYBW-60.F CYBF-250.F CYBW-10Q.NS CYBW-60Q.NS
MSPP-CYBW10QSTCS 684.92 USD InStock
MSPP-CYBW10QST MSPP-CYBF-50ST MSPP-CYBW-60QF MSPP-CYBW60QST MSPP-CYBF250ST MSPP-CYBF-250F MSPP-CYBF-50NS MSPP-CYBW10QNS MSPP-CYBW-60.F MSPP-CYBW-25NS MSPP-CYBW25QST MSPP-CYBW60QNS MSPP-CYBW25QNS MSPP-CYBW-60NS MSPP-CYBW-10ST MSPP-CYBW-25ST MSPP-CYBF-50.F MSPP-CYBW-10NS MSPP-CYBW-25QF MSPP-CYBW-60ST MSPP-CYBW-25.F MSPP-CYBF250NS
Automation Tips for Analytik Jena Cybio Felix, Well Vario and Selma Workstations
Pipette Tips
Fluotics manufactures a selection of automation tips compatible with Analytik Jena Cybio Felix, Well Vario, and Selma Workstations. These automation tips are directly interchangeable with their Beckman automation tip counterpart, with no change to the labware definitions required.
  • Certified RNase-/DNase-free and nonpyrogenic
  • Produced from virgin, high-grade polypropylene
  • Available in non sterile, sterile, and barrier filter formats
Compatible with Analytik Jena Cybio Felix, Well Vario, and Selma Workstations. Available in quadrant and custom fill options. Ultra polishing techniques ensure low liquid retention without the use of releasing agents or other additives.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty