Would you like to log in again?
DecapiCones® Rodent Restrainer, Braintree Scientific
Supplier: Braintree Scientific
DecapiCones®
DC-L120
MSPP-DCL120PK 223.08 USD InStock
MSPP-DCL120
DecapiCones® Rodent Restrainer, Braintree Scientific
Animal Supplies Animal Restraints
Tapered plastic film tubes provide quick and easy restraint of rats, mice, and other small animals.
  • Injections can be made directly through the film
  • Restrain post-decapitation kicking
  • Prevent personal contact with feces or urine
A unique dispenser holds Decapicones open and ready for use.
They may be used repeatedly for injections and simply discarded when soiled. For decapitation, hold at the rear and insert the small end into the decapitator.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty