Would you like to log in again?
Abalone beta-Glucuronidase
Supplier: Campbell Science
DR2102-50 DR2102-25
MSPP-DR2102-50EA 666.34 USD InStock
MSPP-DR2102-50 MSPP-DR210225
Abalone beta-Glucuronidase
Enzymes
beta-Glucuronidase solution is isolated from abalone.
  • >100000 units/ml beta glucuronidase
  • <7000 units/ml sulfatase
  • Low odor and viscosity solution
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty