Would you like to log in again?
ExoQuick Plasma Precipitation Kit, System Biosciences
Supplier: System Biosciences
EXOQ5TM-1
MSPP-EXOQ5TM1EA 1240 USD InStock
MSPP-EXOQ5TM1
ExoQuick Plasma Precipitation Kit, System Biosciences
Assays Cellular Assays
A single kit optimizes exosome isolation from plasma by combining thrombin for de-fibrination with ExoQuick.
  • Saves time and labor
  • Easily scalable
  • Conserves precious sample
  • Delivers high yields of functional, high quality exosomes
Can be used to isolate exosomes for a wide range of downstream applications including biomarker studies exosomal miRNA profiling, exosomal proteomics, exosomal lipidomics/metabolomics, functional studies such as in cell-to-cell signaling, basic biology such as role in tumorigenesis.
Delivery information: The kit includes 5 ml ExoQuick plus 500 µl thrombin at 500 U/ml.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty