Would you like to log in again?
Air Science USA® Laminar Flow Cabinets, Air Science USA LLC
Supplier: Air Science
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FLOW-36-G FLOW-36-A FLOW-48-A FLOW-24-G FLOW-48-G FLOW-24-A
MSPP-FLOW-24-AEA 2127.69 USD InStock
MSPP-FLOW-24-A MSPP-FLOW-36-A MSPP-FLOW-48-A MSPP-FLOW-24-G MSPP-FLOW-36-G MSPP-FLOW-48-G
Air Science USA® Laminar Flow Cabinets, Air Science USA LLC
Fume Hoods and Enclosures Laminar Flow Cabinets
The Purair® FLOW Series vertical laminar flow cabinets are compact and are ideal for use in laboratory environments where space is limited. They are designed to use HEPA filtration to protect equipment and other contents of the work zone from particulates.
  • Vertical laminar airflow with HEPA filtration protects materials inside the cabinet from particulates
  • High efficiency EC Blower
  • Two cable pass through ports located at rear of enclosure
  • Air cleanliness meets ISO Class 5
  • Energy saving LED task lighting
  • Large front opening provides unrestricted access to the work zone
The Purair® FLOW is designed to provide protection to the work surface from biological or particulate contamination. It is ideal for pharmacy compounding of non-toxic agents, assembly of finely tuned instrumentation, cell culture and other processes where protection of the user is not required but easy access to the work surface is important. ISO Class 5 air quality is maintained through a downward laminar flow that provides gentle airflow throughout the cabinet. The clean, simple, low maintenance design offers a comfortable workstation for a range of applications and may be bench mounted or positioned on a mobile cart.
Purair® FLOW Vertical Laminar Flow Workstation, clean bench, with LED task light, 600 mm (24"), 900 mm (36") or 1200 mm (48") Nominal Width, Standard Depth 610 mm (24"), 115 V 60 Hz have North American Cord set and 230 V 50 Hz have UK Cord set (unless specified). Includes main HEPA filter: MSPP-ASTS-030 (1× for 24" and 2× for 48" models) or MSPP-ASVLFP536-030 (1×36" units).
Certifications: UL-C-61010-1, CAN/CSA C22.2 61010-1-12, EN 61010-1:2010. The 230 V units are CE marked.
Delivery information: Units sold complete with all filters included. Optional accessories: Spill tray and base stand for corresponding size cabinet.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty