Would you like to log in again?
Hibernate E Media, BrainBits®
Supplier: BRAINBITS LLC MS
HE
MSPP-HEEA 257.42 USD InStock
MSPP-HE
Hibernate E Media, BrainBits®
Media for Microbiology Prepared Media for Microbiology
Hibernate® is a nutrient medium for the maintenance of neural tissue or cells in ambient carbondioxide levels (US PATENT 6,180,404).
Hibernate-E is used for embryonic tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty