Would you like to log in again?
TOC Vials, Sievers Certified
Supplier: VEOLIA WTS ANALYTICAL INSTR MS
HMI90606
MSPP-HMI90606CS 761.54 USD InStock
MSPP-HMI90606
TOC Vials, Sievers Certified
Vials
Sievers Certified (<10 ppb) TOC Vials are designed for use in critical pharmaceutical applications such as USP and EP water testing as well as municipal water testing. Certification to below 10 ppb gives you confidence in the quality of TOC measurements.
  • Reliable, with guaranteed performance
Scrupulously pre-cleaned in an ISO 9001 environment using a validated, automated process.
Sievers Certified TOC Vials are compatible with most TOC analyzers including Sievers, Shimadzu, OI Analytical, Teledyne-Tekmar and Skalar.
These vials are the right choice for strict regulatory compliance testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty