Would you like to log in again?
HTP-1500 Heat Therapy Pump for Animals, Kent Scientific
Supplier: Kent Scientific
HTP-1500
MSPP-HTP1500EA 944.62 USD InStock
MSPP-HTP1500
HTP-1500 Heat Therapy Pump for Animals, Kent Scientific
Animal Supplies Animal Anesthesia Systems
The HTP-1500 Heat Therapy Pump allows users to safely warm their animals. This is one of the most accurate localized heat therapy system available.
  • Maintain temperature within 1°F for animal safety
  • Mercury-free system decreases potential hazards
  • Uses tap water for added convenience
  • Includes both click-tite and colder connections
  • Accuracy: ± 2 °F
Three temperature safety limits guard against the possibility of overheating and the Hi Limit switches can be tested without taking the unit apart. This provides bright and easy-to-read display which ensures continuous real-time temperature readings.
Temp. set point range: 75 °F to 107 °F (24 °C to 42 °C)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty