Would you like to log in again?
Human VDR Luciferase Reporter Assay Kit
Supplier: INDIGO Biosciences
IB00702 IB00701-32 IB00701
103881-538KT 2830.77 USD InStock
103881-538 MSPP-IB00701 103881-536
Human VDR Luciferase Reporter Assay Kit
Assays Cellular Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Human Vitamin D Receptor reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against VDR (NR1I1).
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
This is an all-inclusive cell-based luciferase reporter assay kit targeting the Human Vitamin D Receptor (VDR). INDIGO's VDR reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the VDR. In addition to VDR Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, Luciferase Detection Reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against human VDR. This kit provides researchers with: Clear, reproducible results, exceptional cell viability post-thaw, consistent results lot to lot.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at –80 °C. Do not store in liquid nitrogen.
Caution: Do not store in liquid nitrogen. Note reporter cells cannot be refrozen or maintained in extended culture.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty