Would you like to log in again?
ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit, Impact Proteomics
Supplier: Impact Proteomics
ProMTag
IP-1000
MSPP-IP-1000EA 448.38 USD InStock
MSPP-IP-1000
ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit, Impact Proteomics
Protein Purification Protein Purification Kits
The ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit produces high-quality antigen samples ready for mass spectrometry analysis.
  • Compatible with a variety of antibody sources including purified antibodies, serum, plasma, bronchoalveolar lavage fluid, and cerebrospinal fluid.
  • Reagents and plasticware for 8 sample preparations are included. User provides protease inhibitors and antibody and antigen sources.
  • Includes Impact Proteomics' MT-Trypsin. The user does not need to provide an enzyme.
  • Requires only standard laboratory equipment.
The ProMTag Immunoprecipitation to Mass Spectrometry (IP-to-MS) Kit produces high-quality antigen samples for mass spectrometry analysis in a fast, simple workflow. Purify target antigens without worrying about contaminating antibodies, detergents, or salts. Each kit contains reagents for 8 immunoprecipitations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty