Would you like to log in again?
Detergent, LancerClean, LCD-P, Lancer
Supplier: Lancer
LancerClean
LCD2.5P
MSPP-LCD25PCS 207.69 USD InStock
MSPP-LCD25P
Detergent, LancerClean, LCD-P, Lancer
Cleaning Agents Detergents and Surface Cleansers
LCD-P is a potassium hydroxide based solution.
Provides excellent cleaning results for all types of contaminants and standard laboratory soils and also it can be used in all glassware washers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty