Would you like to log in again?
MagPlex® Microspheres, Luminex
Supplier: Luminex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MC12005-01 MC12040-01 MC10008-01 MC10070-01 MC12010-01
MSPP-MC1000801EA 1396.92 USD InStock
MSPP-MC1000801 MSPP-MC1204001 MSPP-MC1201001 MSPP-MC1200501 MSPP-MC1007001
MagPlex® Microspheres, Luminex
Beads Magnetic Beads
MagPlex® microspheres are carboxylated polystyrene microparticles, or 'beads' that have been dyed into spectrally distinct sets, or regions allowing them to be individually identified by an xMAP® instrument. These uniquely coded beads provide a user an addressable substrate on which to perform multiple bio-analytical reactions in a single well.
  • Improved washing efficiency
  • Surface carboxyl groups
  • Superparamagnetic
  • Ideal for automated applications
  • No need for expensive filter plates
  • Available in low concentration, for small scale and assay development applications
They are magnetic, and with the use of a magnetic separator, they can be separated from solution quickly and easily. This improves percent recovery during handling and wash steps and facilitates assay automation.
Available in 500 unique regions, enabling the development of assays which can measure up to 500 different analytes in the same well.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty