Would you like to log in again?
Mouse Pie Cage, Braintree Scientific
Supplier: Braintree Scientific
MPC-2
MSPP-MPC2SETEA 746.15 USD InStock
MSPP-MPC2SET
Mouse Pie Cage, Braintree Scientific
Animal Supplies Animal Cages
Mouse pie cages are easy to clean and to use. The circular cages hold up to 11 mice for a whole body irradiation.
  • Notched removable lid can be dialed to any of the ventilated pie-shaped chambers
  • Can also be used for temporary transport or storage of animals for acute studies
Cage has 11 usable sections and one for loading.
Total Dia. 21.5 cm, height: 7.5 cm
Chambers: approx. 5 cm base and 9 cm length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty