Would you like to log in again?
ProMTag Multiomics DNA, RNA, Protein Sample Preparation Kit, Impact Proteomics
Supplier: Impact Proteomics
ProMTag
MULTI-1000
MSPP-MULTI1000EA 475.15 USD InStock
MSPP-MULTI1000
ProMTag Multiomics DNA, RNA, Protein Sample Preparation Kit, Impact Proteomics
Protein Purification Protein Purification Kits
The ProMTag Multiomics DNA, RNA, Protein Sample Preparation kit produces high-quality DNA, RNA, and peptide samples from the same starting sample.
  • Compatible with most starting samples including cultured cells, tissues, and pre-made lysates so long as they do not contain buffers with primary amines (such as TRIS).
  • Reagents and plasticware for 8 sample preparations are included. User provides RNase and/or DNase depending on downstream applications and RNase inhibitors if necessary.
  • Includes Impact Proteomics' MT-Trypsin. The user does not need to provide an enzyme.
  • Requires only standard laboratory equipment.
The ProMTag Multiomics DNA, RNA, Protein Sample Preparation kit utilizes the reversible chemistry of ProMTag to produce high-quality DNA, RNA, and peptide samples from the same starting sample. DNA and RNA are compatible with downstream approaches such as whole genome sequencing and RNA-Seq. Peptides are ready for mass spectrometry analysis. Included reagents and an easy-to-perform workflow take you from cell or tissue lysis all the way to DNA, RNA, and peptides. Each kit contains reagents for 8 sample preparations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty