Would you like to log in again?
Power Supply, P030HP2
Supplier: Acopian Technical Company
P030HP2
MSPP-P030HP2EA 2546.15 USD InStock
MSPP-P030HP2
Power Supply, P030HP2
Power Supplies
High voltage AC-DC rack mounting regulated power supply.
Ideal for laboratory and instrumentation applications, these rack mounting supplies have the same output ratings and specifications as the modular supplies, but additionally feature calibrated ten-turn controls (with locking vernier dials) for precisely setting voltage and current. Voltmeter and ammeter are standard.
Ordering information: With five year warranty.
Delivery information: An 8" long shielded output cable is included.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty