Would you like to log in again?
Glass Bottom Dish/Coverslip, MatTek
Supplier: MatTek
P35G-0-14-C P35G-0-7-C-200 P35G-1.5-10-C
MSPP-P35G1510CCS 583.08 USD InStock
MSPP-P35G1510C MSPP-P35G07C20 MSPP-P35G014C
Glass Bottom Dish/Coverslip, MatTek
Trays/Dishes/Pots Laboratory Trays/Dishes
These glass bottom dishes are designed with the combined technology of the convenient standard size 35 and 50 mm disposable plastic petri dishes with the optical quality of glass to provide superior microscopic images.
  • Sterilized with gamma-irradiation
  • Uncoated
For most applications, No. 1.5 is the preferred coverslip thickness, especially for optimizing image quality with high numerical aperture objectives. The 7 mm diameter microwell is ideal for use with expensive reagents or cell limited applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty