Would you like to log in again?
Automation Tips for PerkinElmer® (Revvity) Janus® MDT and Varispan Workstations
Supplier: Fluotics
PKE-30.NS PKE-200.ST PKE-200.NS PKE-50CB.NS PKE-50.F PKE-235.ST PKE-50.NS PKE-200CB.F PKE-200.F PKE-235.F PKE-50CB.F PKE-50.ST PKE-200CB.ST PKE-235.NS PKE-30.ST PKE-30.F PKE-200CB.NS PKE-50CB.ST
MSPP-PKE-30.NSCS 2130.77 USD InStock
MSPP-PKE-30.NS MSPP-PKE-235.F MSPP-PKE-50.ST MSPP-PKE-30.ST MSPP-PKE-50.NS MSPP-PKE-30.F MSPP-PKE-50.F MSPP-PKE-235ST MSPP-PKE-50CBF MSPP-PKE50CBST MSPP-PKE-235NS MSPP-PKE50CBNS MSPP-PKE20CBNS MSPP-PKE-200NS MSPP-PKE200CBF MSPP-PKE20CBST MSPP-PKE-200ST MSPP-PKE-200.F
Automation Tips for PerkinElmer® (Revvity) Janus® MDT and Varispan Workstations
Pipette Tips
Fluotics manufactures a complete selection of automation tips compatible with the PerkinElmer® (Revvity) Janus® MDT and varispan workstations. These automation tips are directly interchangeable with their PerkinElmer® automation tip counterpart, with no change to the labware definitions required.
  • Certified RNase-/DNase-free and nonpyrogenic
  • Produced from virgin, high-grade polypropylene
  • Available in non-sterile, sterile, and barrier filter formats
Compatible with PerkinElmer® (Revvity) Janus® MDT and varispan workstations. Ultra polishing techniques ensure low liquid retention without the use of releasing agents or other additives.
Packaging: Pipette tips are packaged 96 or 384 per rack, 50 racks per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty