Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PermaLife™ Cell Culture Bags, OriGen

Supplier: OriGen Biomedical
PL120-2G PL325-2G PL30-2G PL70-2G PL240-2G
MSPP-PL120EA 107.69 USD InStock
MSPP-PL120 MSPP-PL70 MSPP-PL240 MSPP-PL30 MSPP-PL325
PermaLife™ Cell Culture Bags, OriGen
Bags Media Bags
PermaLife™ cell culture bags represent the optimal way to harvest, expand, concentrate and freeze critical cell cultures.

  • Contains no plasticizers or chemical residuals
  • Possibility of contamination is less
  • Suitable for cryogenic preservation
  • Temperature resistant from –200 to 200 °C
  • Optical transmission of greater than 95%

Made of FEP, it is inert, so antibodies, cytokines and other genetic material are unaffected by these bags. They are gas permeable and provide a high rate of oxygen transfer to cells under culture. Water permeability is very low, so water loss is negligible and humidified incubators are not required.

Swabable, needle-free valves are provided on every bag to preserve sterile access and reduce the chance of needle sticks. No significant decrease in mechanical strength at temperatures from cryogenic storage (–196 °C) to high autoclave (137 °C).

Delivery information: Supplied with one female luer and one needle-free valve.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty