Would you like to log in again?
QuickZyme Sensitive Tissue Collagen Assay Kit, BioVendor
Supplier: BioVendor
QZBTISCOL5
MSPP-QZBTISCOLEA 2964.8 USD InStock
MSPP-QZBTISCOL
QuickZyme Sensitive Tissue Collagen Assay Kit, BioVendor
Assays Cellular Assays
Collagen is one of the main components of extracellular matrix. Dysregulation in collagen production results in pathologies such as fibrosis (too much collagen) or osteoarthritis (too little collagen). Therefore measurement of collagen production is important in many disease related studies.
  • Quantitative measurement of all types of collagen, species independent
  • Samples: tissue homogenates, tissues
  • Colorimetric measurement
  • Optimized for tissues with low collagen content such as liver
  • No matrix effects as seen in other assays
  • Range: 6 to 300 μg/ml
  • Sensitivity: 2.5 µg/ml
  • Uses rat tail collagen as standard
  • Ease-of-use: Equivalent to ELISA
The QuickZyme sensitive tissue collagen assay is based on the detection of hydroxyproline. Hydroxyproline in tissue hydrolysates can be used as a direct measure of the amount of collagen present in the tissue.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty