Would you like to log in again?
FLIPR® Membrane Potential Assay Kit
Supplier: Molecular Devices
FLIPR®
R8126
MSPP-R8126EA 904.06 USD InStock
MSPP-R8126
FLIPR® Membrane Potential Assay Kit
Assays Cellular Assays
The FLIPR® Membrane Potential Assay Kit provide a fast, simple and reliable, fluorescence-based assay for detecting changes in voltage across the cell membrane and its homogenous fluorescence-based formulations for observation of real-time membrane potential changes associated with ion channel activation and ion transporter proteins.
  • More reproducible data with faster response time
  • No cumbersome pre-assay preparation
  • Ease of use at room to physiological temperatures
  • Fewer steps in the assay resulting in higher sample throughput
Each homµgeneous assay kit utilises a proprietary indicator dye and quencher combination to maximise cell line/channel/compound applicability, while eliminatiµg causes of data variability. This unique formula responds 10 times faster and hasµgreater temperature stability than traditional dyes, providiµg hµgh quality screeniµg data that showsµgood correlation with manual patch clamp assays.
Ion channel activity is highly sensitive and potentially impacted by subtle chemical changes, two FLIPR® Membrane Potential Assay Kits (Red and Blue) are available to select the optimal conditions for your delicate ion channel targets. Both formulations utilise Molecular Devices proprietary quench technology to enhance signal windows and yield acceptable Z-scores to screen a variety of targets, including TRP, ligand-, cyclic nucleotide- and voltage-gated channels.
Delivery information: Explorer kit includes 10 vials of component A and one bottle of component B 1X buffer (1X HBSS + Ca²⁺, Mg²⁺, 20 mM HEPES, pH 7,4). Bulk kit includes 10 vials of component A and one bottle of component B 10X buffer, and Evaluation kit includes 5 vials of membrane potential assay Blue (Component A), 5 vials of membrane potential assay red (component A) and one bottle of component B 1X buffer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty