Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10%, buffered

Supplier: AVANTIK MS
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

RS4061 RS4064
MSPP-RS4061CS 52 USD InStock
MSPP-RS4061 MSPP-RS4064
Formalin 10%, buffered
Formalin
This 10% Neutral Phosphate Buffered Formalin is a ready-to-use fixative.

The cubitainer packaging comes with a convenient carry handle and dispensing spout.


Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty