Would you like to log in again?
MouseTRAP Assay (TRACP 5b Mouse) ELISA, Biovendor
Supplier: BioVendor
SB-TR103
MSPP-SB-TR103EA 1247.89 USD InStock
MSPP-SB-TR103
MouseTRAP Assay (TRACP 5b Mouse) ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of TRAP 5b.
The MouseTRAP™ (TRAcP 5b) ELISA assay uses a polyclonal antibody (7) prepared using recombinant mouse TRACP (11) as antigen. In the test, the antibody is incubated in anti-rabbit IgG coated microtiter wells. After washing, standard, control, and samples are incubated in the wells, and bound TRACP 5b activity is determined with a chromogenic substrate to develop color. The reaction is stopped, and the absorbance of the reaction mixture is read in a microtiter plate reader, color intensity being directly proportional to the amount and activity of TRACP 5b present in the sample.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty