Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Trinocular stereo microscope on ball bearing boom stand
Supplier: UNITED SCOPE LLC DBA AMSCOPE MS
SM-5TZZ
MSPP-SM-5TZZEA 1363.06 USD InStock
MSPP-SM-5TZZ
Trinocular stereo microscope on ball bearing boom stand
Microscopes
It comes with a trinocular head with 30 mm super widefield, high-eyepoint WF10X/20 eyepieces and a ball-bearing double-arm boom stand. This microscope offers high resolution, super widefield of view, large magnifying zoom range and long working distance. It is an ideal instrument for engravers, gemologists, circuit board inspectors and anyone in semiconductor or electronics industry.
  • 3.5-180X zoom microscope on ball-bearing boom stand
  • Ball-bearing stand offering effortless moving adjustments
  • 2- ¹/₂" (65 mm) field view and 8" (200 mm) working distance
  • Trinocular Port for adding photo or video capability
  • Precise ground glass lenses
Both ocular-tubes contains adjustable diopter and rubber eye-guards are included. It provides accurate alignment for ensuring fatigue-free long time viewing.
The heavy-duty cast steel stand finished is with stain resistant enamel and has rugged solid cast steel base.
It provides excellent five year manufacturer warranty.
Certifications: ISO 9001
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty