Would you like to log in again?
Jess or Wes Separation Module, Protein Simple
Supplier: PROTEIN SIMPLE MS
SM-W004
MSPP-SMW004EA 2067.69 USD InStock
MSPP-SMW004
Jess or Wes Separation Module, Protein Simple
Assays Protein Assays
Reagents for 200 data points to use with a detection module.
Includes two parts: Separation box with 8 Capillary Cartridges (25 capillaries per cartridge), 8 Jess/Wes 12-230 kDa Pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 2 and Matrix Removal Buffer), Sample Buffer, Wash Buffer (Store at RT), plus Standard Pack 1 clamshell with 8 ea. DTT, FL Standard master mix with 29 kDa System Control protein. (Store at 4¡C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty