Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

APC Smart-UPS On-Line, 2200VA, Tower, Schneider Electric Systems

Supplier: SCHNEIDER ELECTRIC SYSTEMS MS
APC
SRT2200XLA
MSPP-SRT2200XLEA 2769.23 USD InStock
MSPP-SRT2200XL
APC Smart-UPS On-Line, 2200VA, Tower, Schneider Electric Systems
Power Supplies
High density, double-conversion on-line power protection with scalable runtime.

  • Lead-Acid battery (72 V, 102 W rated, 306 VAh runtime, 3 to 5 year(s) life time) used
  • Audible and visible alarms prioritized by severity
  • Emergency Power Off (EPO) available
  • Plug standard - NEMA 5-20P
  • Power module - 1400
  • Operating altitude: 0 to 10000' and Storage altitude: 0 to 50000' (0.0 to 15240.00 m)

Vertical position, front mounting location and lower preference, not rack-mountable, USB compatible.

Input frequency of 40 to 70 Hz auto-sensing.

Output: Maximum configurable power (Watts) - 1800 W, harmonic distortion - <2%, frequency (sync to mains) 50/60 Hz ±3 Hz sync to mains, Double conversion online topology, crest factor: 3 : 1, Sine wave type, internal bypass (automatic and manual), maximum configurable power - 2200 VA.

Heat dissipation: 535 Btu/h.

IP degree of protection IP20.

Certifications: ENERGY STAR V2.0 (USA), Standards CSA C22.2 No.107.3-05 FCC Part 15 class A UL 1778.

Delivery information: Supplied with Software CD, documentation CD, installation guide, removable support feet, USB cable and warranty card.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty