Would you like to log in again?
Hematology Staining Reagents, Wescor
Supplier: Wescor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SS-035C SS-035/049B SS-035A
MSPP-SS035049BEA 31 USD InStock
MSPP-SS035049B MSPP-SS035C MSPP-SS035A
Hematology Staining Reagents, Wescor
Stains Hematology Stains
Reagents designed for staining peripheral blood smears with the aerospray hematology slide stainer.
  • Shelf life of 18 months
Purpose is to fix granules of basophilic leukocytes in the slide.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty