Would you like to log in again?
CappWash™ Microplate Washer, Pipette.com
Supplier: PIPETTE.COM MS
W-16 W-12 W-8
MSPP-W-12EA 896.54 USD InStock
MSPP-W-12 MSPP-W-8 MSPP-W-16
CappWash™ Microplate Washer, Pipette.com
Microplate Washers
CappWash plate washer is an ideal ELISA plate washer for small-scale work. Simple, compact and inexpensive. Each tube consists of an inner tube that releases the distilled water washing solution into the wells for rinsing while the outer tube vacuums away the liquid upon completion of the washing procedure.
  • Manual ELISA plate washer
  • Simple & robust design
  • Easy to operate
  • Fully autoclavable
  • Highly resistant to chemicals
  • equires no programming nor electrical wiring connections
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty