Would you like to log in again?
Manitowoc Undercounter Flake Ice Machine, Air Cooled
Supplier: Global
WBB3077569
MSPP-WBB307756EA 7030.63 USD InStock
MSPP-WBB307756
Manitowoc Undercounter Flake Ice Machine, Air Cooled
Ice Makers
Manitowoc undercounter flake ice machine comes with built in storage.
  • Stainless steel material
  • Self-contained air cooled condenser
  • Small footprint undercounter unit
  • High load maintenance free bearings for enhanced reliability
  • Heavy-duty stainless steel cabinet resists corrosion
  • Easy access door, slides up and out of the way
Flake ice has many uses including food display, salad bars, healthcare, and cold therapy/healthcare.
Amperage: 5.5 A
Kilowatts per 100 lbs.: 5.05 kW
Ordering information: Three years parts and labor warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty