Would you like to log in again?
ProMTag Intact Protein Universal Protein Cleanup Kit, Impact Proteomics
Supplier: Impact Proteomics
ProMTag
WP-2000
MSPP-WP-2000EA 435 USD InStock
MSPP-WP-2000
ProMTag Intact Protein Universal Protein Cleanup Kit, Impact Proteomics
Protein Purification Protein Purification Kits
The ProMTag Intact Protein Universal Protein Cleanup Kit produces high-quality intact protein samples for mass spectrometry analysis or other downstream applications.
  • Compatible with most starting samples including cultured cells, tissues, biological fluids, and pre-made lysates so long as they do not contain buffers with primary amines (such as TRIS).
  • All reagents and plasticware for 8 sample preparations are included.
  • Requires only standard laboratory equipment.
The ProMTag Intact Protein Universal Protein Cleanup Kit utilizes the reversible chemistry of ProMTag to produce high-quality protein samples for mass spectrometry analysis or other downstream applications in a simple, fast workflow with minimal hands-on time.  Works with any starting sample for rapid,  high yield extraction and cleanup of protein samples. Each kit contains reagents for 8 sample preparations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty