Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
EPA Method 8270 and CLP–Semi-Volatiles (3/90 SOW) Base/Neutrals Surrogates, NSI
Supplier: NSI Lab Solutions
C-376H5
NSC376H5EA 173.33 USD InStock
NSC376H5
EPA Method 8270 and CLP–Semi-Volatiles (3/90 SOW) Base/Neutrals Surrogates, NSI
Standards Environmental Analysis Standards
Mixture is packaged in methylene chloride at a concentration of 5,000µg/ml in an ampule
Mixture contains 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14.
An A2LA traceable standard that has been second source verified in house using an A2LA verified standard.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty