Would you like to log in again?
AccessQuick™ RT-PCR System, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A1703 A1702 A1701
PAA1702EA 775.47 USD InStock
PAA1702 PAA1703 PAA1701
AccessQuick™ RT-PCR System, Promega
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The AccessQuick™ RT-PCR System is an easy and convenient master mix for one-tube RT-PCR.
  • Convenient RT-PCR in One Tube
  • Mix contains Tfl DNA Polymerase, dNTPs, magnesium sulfate and reaction buffer
  • Separate AMV RT provided for important no-RT control reactions
  • Simply combine AccessQuick™ Master Mix with AMV RT, RNA template, primers and water for efficient RT-PCR
The AccessQuick RT-PCR System is an easy and convenient master mix for one-tube RT-PCR. It increases the convenience of performing RT-PCR by combining the following components in a single tube: Tfl DNA Polymerase, dNTPs, magnesium sulfate and reaction buffer. The AMV RT enzyme is provided in a separate tube to allow important no-RT control reactions. The AccessQuick Master Mix is simply added to RNA templates in reaction vials, followed by the AMV RT, primers and water. The AccessQuick RT-PCR Master Mix is intended for routine RT-PCR applications that have been previously optimized and do not require extreme conditions.
Ordering information: Each kit contains AccessQuick™ Master Mix (2X), AMV Reverse Transcriptase and Nuclease-Free Water.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty