Would you like to log in again?
Endotoxin Removal Resin, 100ml, Promega
Supplier: Promega
A2191
PAA2191EA 1045.02 USD InStock
PAA2191
Endotoxin Removal Resin, 100ml, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Wizard MagneSil Tfx System provides a simple method for the rapid isolation of transfection-quality plasmid DNA in a 96-well, high-throughput format.
  • Transfection-Quality Plasmid DNA in 96-Well Format
  • Improved transfection and protein expression results
  • Automated methods available
The Wizard MagneSil Tfx System provides a simple and reliable method for the rapid isolation of transfection-quality plasmid DNA in a 96-well, high-throughput format. The use of MagneSil Paramagnetic Particles for lysate clearing as well as DNA capture circumvents the need for centrifugation or vacuum manifolds, allowing DNA purification with the Wizard MagneSil Tfx System to be completely automated. An automated method has been developed for use of this product with a Beckman Coulter Biomek FX robotic workstation. This procedure requires approximately 45 minutes to process a single 96-well plate. The method can be adapted to other robotic workstations, such as the Beckman Coulter Biomek 2000 or the Tecan Genesis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty