Would you like to log in again?
Integrated Reagent Caps, 4/pk, Promega
Supplier: Promega
A2701
PAA2701PK 16.31 USD InStock
PAA2701
Integrated Reagent Caps, 4/pk, Promega
Closures Bottle Closures
Accessory for use with the ReliaPrep Large Volume HT gDNA System used in automated purification of DNA from 1-10 ml blood on liquid-handling systems.
  • Accessory for use with the ReliaPrep™ Large Volume HT gDNA Isolation System
  • Reagent caps used with reagents for automated purification of DNA from 1-10ml blood on liquid-handling systems.
Reagent bottle for ethanol solution used in automated purification of DNA from 1-10 ml blood on liquid-handling systems. Temperature range: 15 °C to 30 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty