Would you like to log in again?
ImProm-II Reverse Transcriptase, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A3802 A3801 A3803
PAA3803EA 1073.63 USD InStock
PAA3803 PAA3802 PAA3801
ImProm-II Reverse Transcriptase, Promega
Nucleic Acid Reagents RT-PCR and cDNA Reagents
Robust reverse transcriptase. Stable at higher reaction temperatures (37-58 degrees C).
  • Full-Length cDNA Synthesis for the Analysis of Rare or Long Messages
  • Transcription up to 8.9kb, cDNA Synthesis, cDNA cloning
ImProm-II Reverse Transcriptase enables robust, full-length cDNA synthesis for the reproducible analysis of rare or long messages. The ImProm-II Reverse Transcriptase can be used to reverse transcribe total RNA, poly(A)+ mRNA or synthetic transcript RNA templates.
Caution: Storage temperature -30°C to -10°C
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty