Would you like to log in again?
Tissue Lysis Buffer (TLA), Promega
Supplier: Promega Corporation
A5091
PAA5091EA 416.87 USD InStock
PAA5091
Tissue Lysis Buffer (TLA), Promega
Buffers Extraction Buffers
  • Used with the ReliaPrep™ Large Volume HT gDNA Isolation System (Cat.# A2751) or the ReliaPrep™ gDNA Tissue Miniprep Systems (Cat.# A2051, and A2052)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty