Would you like to log in again?
GoTaq Probe qPCR Master Mix, 10ml, Promega®
Supplier: Promega
GoTaq®
A6102
PAA6102EA 752.78 USD InStock
PAA6102
GoTaq Probe qPCR Master Mix, 10ml, Promega®
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
GoTaq Probe qPCR Master Mix is a ready-to-use stabilized 2X formulation containing GoTaq Hot Start Polymerase, MgCl2, dNTPs and proprietary reaction buffer.
  • Convenient qPCR Amplification Using Hydrolysis Detection Probes
  • Ready-to-use mix of optimized components for robust amplification
  • Compatible with both fast and standard cycling methods
  • Sensitive detection for quantitation over a broad dynamic range
The GoTaq Probe qPCR Master Mix is optimized for fast and reproducible quantitative PCR assays in the hydrolysis probe detection format. The master mix is provided as a ready-to-use, stabilized 2X formulation and contains GoTaq Hot Start Polymerase, MgCl2, dNTPs and a proprietary reaction buffer. This master mix does not contain a reference dye; a separate tube of CXR reference dye is included.
Caution: Store all components between –30°C and –10°C. Protect components from light at all times. For best results, mix thawed solutions gently to minimize aeration and foaming, and keep on ice. For short–term storage and frequent use, the GoTaq qPCR Master Mix, 2X, may be kept at 2 to 8°C for up to three months if protected from light.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty