Would you like to log in again?
Maxwell® RSC Blood DNA Kit, Promega
Supplier: Promega
Maxwell®
AS1400
PAAS1400EA 557.91 USD InStock
PAAS1400
Maxwell® RSC Blood DNA Kit, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Maxwell RSC Blood DNA Kit is designed for optimal automated extraction of DNA from up to 300μl of whole blood samples using the Maxwell RSC Instrument
  • Optimized for maximum yield and purity
  • Extracts DNA from 50–250µl of whole blood with a simple, walk-away protocol
  • Provides high yields of pure and amplifiable DNA
  • Compatible with blood stored in EDTA, heparin and citrate anticoagulants
The Maxwell RSC Blood DNA Kit delivers buffy coat-like purity and concentration from whole blood and saves time with automation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty