Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SEV Elution Tubes, 50/pk, Promega

Supplier: Promega
AS5101
PAAS5101EA 73.4 USD InStock
PAAS5101
SEV Elution Tubes, 50/pk, Promega
Tubes Test Tubes
For use with the Maxwell(R) 16 and Maxwell(R) 16 forensic instruments in either SEV or LEV configuration.

  • Standard elution volume (SEV) is 200 to 400 µl
  • Low elution volume (LEV) is 30 to 100 µl

Plungers and elution tubes are only for the Maxwell(R) 16 instruments.

Storage: 15 to 30 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty