Would you like to log in again?
PCR Nucleotide Mix, Promega
Supplier: Promega
C1145 U1432 U1431 C1141
76201-266EA 250.37 USD InStock
76201-266 PAC1145 PAC1141 76201-264
PCR Nucleotide Mix, Promega
Nucleic Acid Reagents Nucleotides
PCR Nucleotide Mix is a premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP, each at a concentration of 10mM or 25mM in water.
  • cGMP-Grade Solution of dNTPs Pretested in PCR
  • cGMP-manufactured for lot-to-lot product consistency
  • Equimolar amounts of each dNTP in water
  • Two concentrations available: 10mM and 25mM
High-quality deoxynucleotide triphosphates (dNTPs) are critical for PCR success. PCR Nucleotide Mix is a cGMP-grade premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP. There are two formulations available: A premixed solution containing each nucleotide at a concentration of 10mM in water at pH 7.5; the total concentration of nucleotides is 40mM. A premixed solution containing each nucleotide at a concentration of 25mM in water at pH 7.5; the total concentration of nucleotides is 100mM. These PCR nucleotide mixes are ready to use and optimized for use in PCR.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty