Would you like to log in again?
T4 Polynucleotide Kinase Buffer Pack
Supplier: Promega
C1313
PAC1313EA 39.98 USD InStock
PAC1313
T4 Polynucleotide Kinase Buffer Pack
Buffers Cloning Buffers
Designed for use with T4 Polynucleotide Kinase to catalyze the transfer of the γ-phosphate from ATP to the 5´-terminus of poly- or mononucleotides bearing a 5´-hydroxyl group.
  • Use with T4 Polynucleotide Kinase to phosphorylate RNA and DNA
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty