Would you like to log in again?
CXR Reference Dye, Promega
Supplier: Promega
C5411
PAC5411EA 34.52 USD InStock
PAC5411
CXR Reference Dye, Promega
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Use Carboxy-X-Rhodamine (CXR) Reference Dye (identical to ROX™ dye) with instruments that require a higher level of reference dye that than in the GoTaq® qPCR Master Mix. Component of GoTaq® qPCR and RT-qPCR Systems.
  • Use with instruments that require a higher level of reference dye
  • Component of GoTaq® qPCR and RT-qPCR Systems
  • Excitation at 580nm and emission at 602nm
Caution: Storage temperature: -30°C to -10°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty