Would you like to log in again?
psiCHECK-2 Vector, 20 µg, Promega
Supplier: Promega
C8021
PAC8021EA 526.87 USD InStock
PAC8021
psiCHECK-2 Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
psiCHECK-1 and psiCHECK-2 Vectors enable monitoring of changes in expression of a target gene fused to a reporter gene. They are used for optimizing RNA interference assays.
  • Vectors for Optimizing RNA Interference Assays
  • Monitor changes in expression of a target gene fused to a reporter gene; decreases in luciferase activity correlate with RNAi
  • psiCHECK™-1 Vector is used to monitor RNAi effects in live cells
  • psiCHECK™-2 Vector designed for endpoint lytic assays
The psiCHECK-1 and psiCHECK-2 Vectors are designed to provide a quantitative and rapid approach for initial optimization of RNA interference (RNAi). The vectors enable monitoring of changes in expression of a target gene fused to a reporter gene. In both vectors Renilla luciferase is used as the primary reporter gene, and the gene of interest is cloned into a multiple cloning region located downstream of the Renilla translational stop codon. Initiation of the RNAi process by synthetic siRNAs toward a gene of interest results in cleavage and subsequent degradation of the fusion mRNA. Measuring decreases in Renilla activity provides a convenient way of monitoring the RNAi effect. In comparison with other fusion approaches (e.g., GFP or flag-tags), the Renilla luciferase approach offers more convenient and rapid quantitation with higher sensitivity. The psiCHECK-1 Vector is recommended for use in monitoring RNAi effects in live cells. The changes in Renilla luciferase activity are measured with the EnduRen Live Cell Substrate (Cat.# E6481), which allows continuous monitoring of intracellular Renilla luminescence. The psiCHECK-2 Vector contains a second reporter gene, firefly luciferase, and is designed for endpoint lytic assays. Introduction of firefly luciferase in the psiCHECK-2 Vector allows normalization of Renilla luciferase expression, achieving robust and reproducible results.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty