Would you like to log in again?
Dual-Luciferase Reporter 1000 Assay System, 1,000 assays, Promega
Supplier: Promega
E1960 E1980 E1910
PAE1980EA 1693.19 USD InStock
PAE1980 PAE1960 PAE1910
Dual-Luciferase Reporter 1000 Assay System, 1,000 assays, Promega
Assays Enzyme Assays
The Dual-Luciferase Reporter (DLR) Assay System provides an efficient means of performing two reporter assays. In the DLR Assay, the activities of firefly and Renilla luciferases are measured sequentially from a single sample.
  • Detect firefly and Renilla Luciferase Activities in a Single Sample
  • Renilla luciferase internal control provides more accurate results
  • Range extends over 7 logs; very active samples typically do not need dilution
  • Requires luminometer with 2 injectors
The Dual-Luciferase Reporter (DLR) Assay System provides an efficient means of performing two reporter assays. In the DLR Assay, the activities of firefly (Photinus pyralis) and Renilla (Renilla reniformis or sea pansy) luciferases are measured sequentially from a single sample. The firefly luciferase reporter is measured first by adding Luciferase Assay Reagent II (LAR II) to generate a luminescent signal lasting at least one minute. After quantifying the firefly luminescence, this reaction is quenched, and the Renilla luciferase reaction is initiated simultaneously by adding Stop & Glo Reagent to the same sample. Both assays can be completed in about 4 seconds using a luminometer with reagent auto-injectors. In the DLR Assay System, both reporters yield linear assays with attomole (100ul/well), additional Passive Lysis Buffer may be purchased separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty