Would you like to log in again?
Steady-Glo Luciferase Assay System, Promega
Supplier: Promega
E2550 E2520 E2510
PAE2520EA 1049.29 USD InStock
PAE2520 PAE2550-A PAE2510
Steady-Glo Luciferase Assay System, Promega
Assays Enzyme Assays
Steady-Glo Luciferase Assay System is a homogeneous assay with signal half-life of over 5 hours in commonly used cell culture media. Ideal for batch processing.
  • Long-Lived Luminescence for Batch Processing in Multiwell Plates
  • Stable luminescence signal with 5-hour half-life
  • No need to remove culture medium or wash cells prior to adding assay reagent
  • Less sensitive to mixing conditions in multiwell plates. Particularly useful in 384-well plates
High-throughput quantitation of firefly (Photinus pyralis) luciferase expression in mammalian cells is commonly performed by batch processing of 96- and 384-well plates. Steady-Glo Luciferase Assay System is designed for this purpose by providing long-lived luminescence when added to cultured cells. The homogeneous assay provides signal half-lives of over 5 hours in commonly used cell culture media without prior sample processing. Throughput rates of several thousand samples per hour may be achieved with high reproducibility under standard laboratory conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty