Would you like to log in again?
QuantiFluor® dsDNA System, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
E2671 E2670
PAE2670EA 374.99 USD InStock
PAE2670 PAE2671
QuantiFluor® dsDNA System, Promega
Assays qPCR Assays
The QuantiFluor dsDNA System contains a fluorescent DNA-binding dye that enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA) in solution. The dye shows minimal binding to single stranded DNA (ssDNA) and RNA.
  • Sensitive Quantitation of Double-Stranded DNA in Solution
  • Specific quantitation of dsDNA
  • More sensitive than NanoDrop for low-concentration samples
The QuantiFluor dsDNA System contains a fluorescent DNA-binding dye that enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA) in solution. The quantitation of dsDNA is a very important step in many biological applications, particularly in standard molecular biology techniques. The dye shows minimal binding to single-stranded DNA (ssDNA) and RNA, allowing specific quantitation of dsDNA.
The dye shows minimal binding to single-stranded DNA (ssDNA) and RNA, allowing specific quantitation of dsDNA.
Caution: Storage conditions: Product may arrive frozen. Upon receipt, store the QuantiFluor® dsDNA Dye at 2–10°C, protected from light. Store the Lambda DNA Standard, 100μg/ml, at 2–10°C. Do not refreeze the Lambda DNA Standard, 100µg/ml. Store the 20X TE Buffer at –30°C to +30°C.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty