Would you like to log in again?
Renilla-Glo Luciferase Assay System, Promega
Supplier: Promega
E2720 E2710
PAE2720EA 1150.93 USD InStock
PAE2720 PAE2710
Renilla-Glo Luciferase Assay System, Promega
Assays Enzyme Assays
The Renilla-Glo Luciferase Assay System is a single-addition reagent that generates a glow-type signal with Renilla luciferase.
  • Simple "Add-and-Read" Renilla Luciferase Reporter Assay
  • No need for separate lysis and reagent injection steps
  • Extended bright light output optimized for batch and continuous-processing
  • Low background provides increased sensitivity
The Renilla-Glo Luciferase Assay System is a single-addition reagent that generates a glow-type signal with Renilla luciferase. When reconstituted, it has the capacity to lyse cells, reduce the autoluminescence of the coelenterazine substrate, and produce a stable signal (i.e., half-life greater than 60 minutes at 22°C).
This system can be used for transcription regulation, virus-cell interactions, compound screening, and more.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty