Would you like to log in again?
Renilla Luciferase Assay System, Promega
Supplier: Promega
E2810 E2820
PAE2810PK 201.63 USD InStock
PAE2810 PAE2820
Renilla Luciferase Assay System, Promega
Assays Enzyme Assays
Renilla Luciferase Assay System provides a fast, sensitive method for detecting luciferase from sea pansy (Renilla reniformis).
  • Fast, Sensitive Detection of Renilla Luciferase Activity
  • Linear range extends over 7 logs
  • Low background, increased sensitivity
  • Detect 10-19 moles of Renilla luciferase
Renilla Luciferase Assay System is designed to provide a fast and sensitive method of detecting luciferase from sea pansy (Renilla reniformis). The system is a convenient alternative to firefly (Photinus pyralis) reporter systems and is designed to yield reliable, linear results for a concentration range over 7 orders of magnitude. The Renilla Luciferase Assay System has been formulated with a proprietary composition that significantly reduces the effect of coelenterazine autoluminescence when compared to other reagents, making the reagent orders of magnitude more sensitive than published methods. This system enables measurements with wildtype and synthetic hRluc genes for primary expression or internal normalization measurements of gene expression.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty