Would you like to log in again?
Beta-Glo Assay System, Promega
Supplier: Promega
E4780 E4720 E4740
PAE4740EA 1151.1 USD InStock
PAE4740 PAE4720 PAE4780
Beta-Glo Assay System, Promega
Assays Enzyme Assays
The Beta-Glo Assay System is a homogeneous, luminescence-based method for quantitating beta-galactosidase expression in mammalian cells.
  • Luminescence Detection of β-Gal Activity
  • Add a single reagent directly to cells in growth medium
  • Optimal assay performance at room temperature
  • Read results using any luminometer. Injectors not required
The Beta-Glo Assay System is a homogeneous method of quantitating beta-galactosidase expression in mammalian cells. The system provides a bright luminescent signal that is stable over several hours in commonly used cell culture medium without prior sample processing. The homogeneous assay procedure involves the addition of a single reagent directly to cells cultured in serum-supplemented medium. Throughput rates of several thousand samples per hour can be achieved with high reproducibility under standard laboratory conditions.
The homogeneous assay procedure involves the addition of a single reagent directly to cells cultured in serum-supplemented medium. Throughput rates of several thousand samples per hour can be achieved with high reproducibility under standard laboratory conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty