Would you like to log in again?
QuantiFluor ONE dsDNA System, Promega
Supplier: Promega
QuantiFluor®
E4870 E4871
PAE4871EA 117.75 USD InStock
PAE4871 PAE4870
QuantiFluor ONE dsDNA System, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The QuantiFluor(R) ONE dsDNA System enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA). The system contains a fluorescent dsDNA-binding dye (504nm[Ex]/531nm[Em]) developed for use in an “add-and-read” format for dye and standard.
  • Fluorescent Quantitation of dsDNA with "Add-and-Read" Convenience
  • Specific quantitation of dsDNA
  • Increased sensitivity over absorbance at 260nm for low-concentration samples
  • Simple to use and automation-friendly
The QuantiFluor ONE dsDNA System contains a fluorescent double-stranded DNA-binding dye (504nmEx/531nmEm) developed for use in an “add-and-read” format for dye and standard, making sample quantitation easy. This system enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA). The QuantiFluor ONE dsDNA System was developed using the fluorescence module of the GloMax Multi+ Detection System with Instinct Software, GloMax Discover System and the Quantus Fluorometer. The QuantiFluor ONE dsDNA System can be used with any fluorometer that is capable of measuring fluorescence at the appropriate excitation and emission wavelengths.
The QuantiFluor ONE dsDNA System can be used with any fluorometer that is capable of measuring fluorescence at the appropriate excitation and emission wavelengths. This system contains sufficient reagents for 100 or 500 assays at 200 μL scale.
The QuantiFluor ONE dsDNA System was developed using the fluorescence module of the GloMax® Multi+ Detection System with Instinct Software, GloMax Discover System, and the Quantus Fluorometer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty