Would you like to log in again?
rCTP, rATP, rUTP, rGTP, 100 mM each, 4×400 microl, Promega
Supplier: Promega
E6000
PAE6000EA 348.83 USD InStock
PAE6000
rCTP, rATP, rUTP, rGTP, 100 mM each, 4×400 microl, Promega
Nucleic Acid Reagents Nucleotides
High-quality rNTPs. Set of four provided at concentrations of 10mM or 100mM in water at neutral pH.
  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • Provided at 10mM (4 x 0.5ml) or 100mM (4 x 400µl) in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC
Set of four rNTPs (rATP, rUTP, rCTP, rGTP) provided in individual tubes and qualified for use with the Riboprobe and HeLaScribe Systems. The rNTPs are supplied in nuclease-free water. Purity is verified by HPLC analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty